missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.

Product Code. 10259470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10259470 1 g Glass bottle 1g
10340344 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259470 Supplier Thermo Scientific Acros Supplier No. 424750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79322-76-2 | C10H12O3 | 180.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79322-76-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00216476
InChI Key KAXLTAULVFFCNL-UHFFFAOYSA-N
PubChem CID 586417
IUPAC Name methyl 4-(1-hydroxyethyl)benzoate
SMILES CC(C1=CC=C(C=C1)C(=O)OC)O

Specifications

Color Colorless to Yellow
Boiling Point 148°C to 152°C (1.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.536
Quantity 1 g
Synonym methyl 4-1-hydroxyethyl benzoate, 4-1-hydroxyethyl benzoic acid methyl ester, 4-1-hydroxy-ethyl-benzoic acid methyl ester, benzoic acid, 4-1-hydroxyethyl-, methyl ester, acmc-20m5ni, benzoic acid, 4-1s-1-hydroxyethyl-, methyl ester, methyl 4-hydroxyethyl benzoate, methyl4-1-hydroxyethyl benzoate, methyl-4-1-hydroxyethyl benzoate, methyl 4-1-hydroxyethyl benzoate #
Solubility Information Solubility in water: slightly
Formula Weight 180.2
Percent Purity 90%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Methyl 4-(1-hydroxyethyl)benzoate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.