missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trifluoromethyl)benzoate, 97%

Product Code. 11486857
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486857 25 g 25g
11476857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486857 Supplier Thermo Scientific Alfa Aesar Supplier No. A14316.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2557-13-3 | C9H7F3O2 | 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2557-13-3
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00043543
InChI Key QQHNNQCWKYFNAC-UHFFFAOYSA-N
Synonym methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride
PubChem CID 520213
IUPAC Name methyl 3-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Density 1.295
Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.453
Quantity 25 g
Beilstein 1963428
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material Methyl 3-(trifluoromethyl)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.