missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxotetrahydrothiophene-2-carboxylate, 98%, Thermo Scientific™

Catalog Number p-7058941
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2689-69-2
C6H8O3S
160.187
MFCD00156156
TUSSXVYKOOVOID-UHFFFAOYSA-N
methyl 3-oxotetrahydrothiophene-2-carboxylate, 2-thiophenecarboxylic acid, tetrahydro-3-oxo-, methyl ester, methyl tetrahydro-3-oxo-2-thenoate, methyl 3-oxo-2,3,4,5-tetrahydrothiophene-2-carboxylate, acmc-1cmcz, 2-methoxycarbonyl-3-oxotetrahydrothiophene, methyl 3-oxo-tetrahydrothiophene-2-carboxylate, 2-carbomethoxy-2,3,4,5-tetrahydrothiophen-3-one, methyl 3-oxo-2,4,5-trihydrothiophene-2-carboxylate
102870
methyl 3-oxothiolane-2-carboxylate
COC(=O)C1C(=O)CCS1
This item is not returnable. View return policy
2689-69-2
C6H8O3S
160.187
MFCD00156156
TUSSXVYKOOVOID-UHFFFAOYSA-N
methyl 3-oxotetrahydrothiophene-2-carboxylate, 2-thiophenecarboxylic acid, tetrahydro-3-oxo-, methyl ester, methyl tetrahydro-3-oxo-2-thenoate, methyl 3-oxo-2,3,4,5-tetrahydrothiophene-2-carboxylate, acmc-1cmcz, 2-methoxycarbonyl-3-oxotetrahydrothiophene, methyl 3-oxo-tetrahydrothiophene-2-carboxylate, 2-carbomethoxy-2,3,4,5-tetrahydrothiophen-3-one, methyl 3-oxo-2,4,5-trihydrothiophene-2-carboxylate
102870
methyl 3-oxothiolane-2-carboxylate
COC(=O)C1C(=O)CCS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 160.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-oxotetrahydrothiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'