missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Oxo-desoxycholate, TRC

Product Code. 30474133
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474133 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474133 Supplier Toronto Research Chemicals Supplier No. TRCM3212002.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-Oxo-desoxycholate
CAS 10538-58-6
Molecular Formula C25H40O4
Quantity 2.5 g
Synonym 12-Hydroxy-3-oxo-cholan-24-oic Acid Methyl Ester, Methyl 12alpha-Hydroxy-3-oxo-5beta-cholanate, Methyl-3-keto-12alpha-hydroxy-5beta-cholanoate
InChI Formula InChI=1S/C25H40O4/c1-15(5-10-23(28)29-4)19-8-9-20-18-7-6-16-13-17(26)11-12-24(16,2)21(18)14-22(27)25(19,20)3/h15-16,18-22,27H,5-14H2,1-4H3/t15-,16-,18+,19-,20+,21+,22+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name methyl (4R)-4-[(5R,8R,9S,10S,12S,13R,14S,17R)-12-hydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 404.58
Formula Weight 404.2927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.