missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Nitroisonicotinate, TRC

Product Code. 30240956
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240956 500 mg 500mg
30240555 100 mg 100mg
30240605 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240956 Supplier Toronto Research Chemicals Supplier No. TRCM340050500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103698-10-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.13
Synonym Methyl 3-Nitropyridine-4-carboxylate;
IUPAC Name methyl 3-nitropyridine-4-carboxylate
SMILES COC(=O)c1ccncc1[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H6N2O4/c1-13-7(10)5-2-3-8-4-6(5)9(11)12/h2-4H,1H3
Formula Weight 182.0328
Chemical Name or Material Methyl 3-Nitroisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.