missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(methylthio)propionate, 98%

Product Code. 11413394
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413394 100 g 100g
11403394 25 g 25g
11423394 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413394 Supplier Thermo Scientific Alfa Aesar Supplier No. A10649.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13532-18-8 | C5H10O2S | 134.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13532-18-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00008565
InChI Key DMMJVMYCBULSIS-UHFFFAOYSA-N
Synonym methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate
PubChem CID 61641
IUPAC Name methyl 3-methylsulfanylpropanoate
SMILES COC(=O)CCSC

Specifications

Density 1.074
Boiling Point 72°C to 73°C (11 mmHg)
Flash Point 72°C (161°F)
Linear Formula CH3SCH2CH2CO2CH3
Refractive Index 1.465
Quantity 100 g
Beilstein 1745077
Formula Weight 134.2
Percent Purity 98%
Chemical Name or Material Methyl 3- (methylthio)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.