missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylphenylacetate, 98%

Product Code. 10650745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10650745 5 g Glass bottle 5g
10650935 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10650745 Supplier Thermo Scientific Acros Supplier No. 374080050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53088-69-0 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53088-69-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00968468
InChI Key AWTKFTNNPQGGLX-UHFFFAOYSA-N
Synonym methyl 2-m-tolyl acetate, methyl 3-methylphenylacetate, methyl 2-3-methylphenyl acetate, 3-methylphenylacetic acid methyl ester, benzeneacetic acid, 3-methyl-, methyl ester, methyl 2-m-tolylacetate, m-tolylacetic acid methyl ester, m-toly-acetic acid methyl ester, m-tolyl-acetic acid methyl ester, methyl 2-3-methylphenyl ethanoate
PubChem CID 5135288
IUPAC Name methyl 2-(3-methylphenyl)acetate
SMILES COC(=O)CC1=CC=CC(C)=C1

Specifications

Color Colorless to Yellow
Boiling Point 117°C (13.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2COOCH3
Refractive Index 1.5045 to 1.5065
Quantity 5 g
Formula Weight 164.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-methylphenylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.