missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate, Mikromol
Description
Methyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate, Mikromol
Specifications
Specifications
| Chemical Name or Material | 3-Methylflavone-8-carboxylic acid methyl ester |
| CAS | 90101-87-4 |
| Analyte or Component Names | 3-Methylflavone-8-carboxylic acid methyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H14 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate, 3-Methyl-flavone-8-carboxylic Acid Methyl Ester |
| InChI Formula | InChI=1S/C18H14O4/c1-11-15(19)13-9-6-10-14(18(20)21-2)17(13)22-16(11)12-7-4-3-5-8-12/h3-10H,1-2H3 |
| Solution Type | Neat |
| SMILES | COC(=O)c1cccc2C(=O)C(=C(Oc12)c3ccccc3)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?