missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methyl-2-furoate, 98%

Product Code. 11498813
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498813 5 g 5g
11408820 10 g 10g
11408823 25 g 25g
11418823 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498813 Supplier Thermo Scientific Alfa Aesar Supplier No. A10541.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6141-57-7 | C7H8O3 | 140.14 g/mol

Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

Solubility
Insoluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6141-57-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00014110
InChI Key AQQYRDKMXXSIMP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester
PubChem CID 350231
IUPAC Name methyl 3-methylfuran-2-carboxylate
SMILES COC(=O)C1=C(C)C=CO1

Specifications

Melting Point 33°C to 37°C
Boiling Point 60°C (1 mmHg)
Flash Point 82°C (180°F)
Quantity 5 g
Beilstein 116835
Solubility Information Insoluble in water.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Methyl 3-methyl-2-furoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.