missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxyphenylacetate, 97%

Product Code. p-7024961
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469706 2.5 g 2.5g
11479706 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469706 Supplier Thermo Scientific Alfa Aesar Supplier No. B20237.88
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18927-05-4 | C10H12O3 | 180.203 g/mol

Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18927-05-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00017205
InChI Key BSVIOYCZTJRBDB-UHFFFAOYSA-N
Synonym methyl 2-3-methoxyphenyl acetate, methyl 3-methoxyphenylacetate, benzeneacetic acid, 3-methoxy-, methyl ester, acetic acid, m-methoxyphenyl-, methyl ester, methyl 3-methoxyphenyl acetate, 3-methoxy-phenyl-acetic acid methyl ester, acmc-1brer, methyl m-methoxyphenylacetate, methyl 3-methoxy-phenylacetate
PubChem CID 519609
IUPAC Name methyl 2-(3-methoxyphenyl)acetate
SMILES COC1=CC=CC(=C1)CC(=O)OC

Specifications

Boiling Point 86°C to 88°C (0.5 mmHg)
Quantity 2.5 g
Solubility Information Sparingly soluble in water.
Formula Weight 180.23
Percent Purity 97%
Chemical Name or Material Methyl 3-methoxyphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.