missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxyacrylate, 95%

Product Code. 10622892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10622892 100 mL Glass bottle 100mL
10376591 25 mL Glass bottle 25mL
10495115 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10622892 Supplier Thermo Scientific Acros Supplier No. 330461000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34846-90-7 | C5H8O3 | 116.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34846-90-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.13
MDL Number MFCD00128206
InChI Key AUTCCPQKLPMHDN-ONEGZZNKSA-N
Synonym methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
PubChem CID 5323651
IUPAC Name methyl (E)-3-methoxyprop-2-enoate
SMILES COC=CC(=O)OC

Specifications

Melting Point 3.0°C to 4.0°C
Color Colorless
Density 1.0700g/mL
Boiling Point 169.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H3COCH=CHCOOCH3
Refractive Index 1.4490 to 1.4510
Quantity 100 mL
Specific Gravity 1.07
Viscosity 1.68 mPa.s (20°C)
Formula Weight 116.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 3-methoxyacrylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3-methoxyacrylate
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.