missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxy-2-nitrobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11372766
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11372766 Supplier Thermo Scientific Alfa Aesar Supplier No. L08886.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

Methyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5307-17-5
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00051968
InChI Key FDQQRLPHAAICCR-UHFFFAOYSA-N
Synonym benzoic acid, 3-methoxy-2-nitro-, methyl ester, 3-methoxy-2-nitrobenzoic acid methyl ester, m-anisic acid, 2-nitro-, methyl ester, maybridge1_002124, cbmicro_008502, methyl 2-nitro-m-anisate, methyl 3-methoxy-2-nitro-benzoate, 2-nitro-m-anisic acid methyl ester, 3-methoxy-2-nitro-benzoic acid methyl ester
PubChem CID 79195
IUPAC Name methyl 3-methoxy-2-nitrobenzoate
SMILES COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O

Specifications

Melting Point 142°C to 144°C
Quantity 5 g
Beilstein 2697098
Solubility Information Insoluble in water.
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxy-2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.