missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Mercaptopropionate, TRC

Product Code. 30464413
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464413 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464413 Supplier Toronto Research Chemicals Supplier No. TRCM32026510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-Mercaptopropionate
CAS 2935-90-2
Molecular Formula C4H8O2S
Quantity 10 g
Synonym 3-Mercaptopropanoic Acid Methyl Ester, 3-Mercaptopropionic Acid Methyl Ester, 3-Mercaptopropanoic Acid Methyl Ester, 3-Mercaptopropionic Acid Methyl Ester, 3-Mercatopropropanoic Acid Methyl Ester, 3-Thiopropanoic Acid Methyl Ester, MPM, MPM (thiol), Methyl 3-Mercaptopropionoate, Methyl beta-Mercaptopropionate, NSC 137814, beta-Mercaptopropionic Acid Methyl Ester
InChI Formula InChI=1S/C4H8O2S/c1-6-4(5)2-3-7/h7H,2-3H2,1H3
SMILES COC(=O)CCS
IUPAC Name methyl 3-sulfanylpropanoate
Molecular Weight (g/mol) 120.17
Formula Weight 120.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.