missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-iodo-4-methylbenzoate, 98%

Product Code. 11492120
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492120 5 g 5g
11402130 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492120 Supplier Thermo Scientific Alfa Aesar Supplier No. H28734.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90347-66-3 | C9H9IO2 | 276.073 g/mol

Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-Iodo-4-methylbenzoate is used as a reagent in the synthesis of [(pyrazolo[1,5-a]pyridmidinyl)ethynyl]benzamides as selective discoidin domain receptor 1 (DDR1) inhibitors. Also used as a reagent in the preparation of orally bioavailable GZD824 targeting Breakpoint Cluster Region-Abelson (Bcr-Abl) kinase and overcoming antitumor drug resistance.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90347-66-3
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00230583
InChI Key NKMHAOTZPFVSPC-UHFFFAOYSA-N
Synonym 3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r
PubChem CID 13614033
IUPAC Name methyl 3-iodo-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)I

Specifications

Refractive Index 1.5975
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 276.07
Percent Purity 98%
Chemical Name or Material Methyl 3-iodo-4-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.