missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxybenzoate, 99%

Product Code. 11457067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457067 500 g 500g
11437067 25 g 25g
11447067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457067 Supplier Thermo Scientific Alfa Aesar Supplier No. A14425.36
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19438-10-9 | C8H8O3 | 152.15 g/mol

Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

Solubility
Soluble in chloroform, dimetyl Sulfoxide,ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19438-10-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002295
InChI Key YKUCHDXIBAQWSF-UHFFFAOYSA-N
Synonym methyl m-hydroxybenzoate, 3-hydroxybenzoic acid methyl ester, m-carbomethoxyphenol, methyl m-oxybenzoate, m-hydroxybenzoic acid methyl ester, benzoic acid, 3-hydroxy-, methyl ester, 3-hydroxy-benzoic acid methyl ester, unii-569m9g3stj, 3-methoxycarbonyl phenol, benzoic acid, m-hydroxy-, methyl ester
PubChem CID 88068
IUPAC Name methyl 3-hydroxybenzoate
SMILES COC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 70°C to 72°C
Boiling Point 280°C to 281°C (708 mmHg)
Quantity 500 g
Beilstein 2208129
Solubility Information Soluble in chloroform,dimetyl Sulfoxide,ethanol and methanol.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Methyl 3-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.