missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-methoxybenzoate, 98%

Product Code. 10103294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10103294 5 g 5g
10505914 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103294 Supplier Thermo Scientific Acros Supplier No. 399260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6702-50-7 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6702-50-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD01321262
InChI Key QXOXUEFXRSIYSW-UHFFFAOYSA-N
Synonym methyl isovanillate, methyl 3-hydroxy-4-methoxybenzenecarboxylate, 3-hydroxy-4-methoxybenzoic acid methyl ester, isovanillic acid methyl ester, methyl-3-hydroxy-4-methoxybenzoate, methyl 3-hydroxy-4-methoxy-benzoate, benzoic acid, 3-hydroxy-4-methoxy-, methyl ester, methyl 3-hydroxy-4-methoxy benzoate, isovamllic acid methyl ester, ksc496e5l
PubChem CID 4056967
IUPAC Name methyl 3-hydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC(O)=C(OC)C=C1

Specifications

Melting Point 64.0°C to 67.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)COOCH3
Quantity 5 g
Formula Weight 182.17
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-methoxybenzoate
compliance-icons
Product Identifier
  • Methyl 3-hydroxy-4-methoxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.