missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-dimethylamino-2-methylpropionate, 98%

Product Code. 11449336
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11449336

Brand: Thermo Scientific Alfa Aesar A19034.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 10205-34-2 | C7H16NO2 | 146.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10205-34-2
Molecular Formula C7H16NO2
Molecular Weight (g/mol) 146.21
MDL Number MFCD00044842
InChI Key DDLJXUCYCFDNCA-ZCFIWIBFSA-O
PubChem CID 112023
SMILES COC(=O)[C@H](C)C[NH+](C)C

Specifications

Boiling Point 152°C to 154°C
Refractive Index 1.419
Quantity 50 g
Beilstein 1755266
Synonym methyl 3-dimethylamino-2-methylpropanoate, methyl 3-dimethylamino-2-methylpropionate, methyl .beta.-dimethylaminoisobutyrate, methyl beta-dimethylamino isobutyrate, 3-dimethylamino-2-methylpropionic acid methyl ester, 3-dimethylamino-2-methyl-propionic acid methyl ester, propanoic acid,3-dimethylamino-2-methyl-,methyl ester, propanoic acid, 3-dimethylamino-2-methyl-, methyl ester, propanoic acid,3-dimethylamino-2-methyl-, methyl ester
IUPAC Name methyl 3-(dimethylamino)-2-methylpropanoate
Formula Weight 145.2
Percent Purity 98%
Chemical Name or Material Methyl 3-dimethylamino-2-methylpropionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.