missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyano-4-methoxybenzoate, 97%

Product Code. p-7045547
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11307566

Brand: Thermo Scientific Alfa Aesar L11107.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25978-74-9 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25978-74-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00052930
InChI Key RYJSFYBJYKFNCF-UHFFFAOYSA-N
Synonym benzoic acid, 3-cyano-4-methoxy-, methyl ester, acmc-1cj4t, methyl 3-cyano-4-methoxy-benzoate, methyl 3-cyano-4-methoxybenzoate., methyl 3-cyano-4-methyloxy benzoate, 3-cyano-4-methoxy-benzoic acid methyl ester, benzoicacid,3-cyano-4-methoxy-,methyl ester
PubChem CID 2801011
IUPAC Name methyl 3-cyano-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1)C(=O)OC)C#N

Specifications

Melting Point 123°C to 127°C
Quantity 1 g
UN Number UN3439
Beilstein 2374769
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material Methyl 3-cyano-4-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.