missing translation for 'onlineSavingsMsg'
Learn More

methyl 3-chlorosulfonylthiophene-2-carboxylate, Thermo Scientific™

Product Code. 10713157
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713157 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10713157 Supplier Thermo Scientific Maybridge Supplier No. GK00438DE
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 59337-92-7
Packaging Amber Glass Bottle
Molecular Formula C6H5ClO4S2
MDL Number MFCD00068160
Quantity 5 g
Synonym methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
InChI Key PJVJBDAUWILEOG-UHFFFAOYSA-N
SMILES COC(=O)C1=C(C=CS1)S(Cl)(=O)=O
IUPAC Name methyl 3-chlorosulfonylthiophene-2-carboxylate
Molecular Weight (g/mol) 240.67
PubChem CID 2736733
Formula Weight 240.68
Percent Purity 95%
Chemical Name or Material methyl 3-chlorosulfonylthiophene-2-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.