missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(chlorosulfonyl)-4-methoxybenzoate, 97%, Thermo Scientific™

Product Code. p-2705217
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10181122

Brand: Thermo Scientific Maybridge MO00348DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 192323-12-9
Molecular Formula C9H9ClO5S
Molecular Weight (g/mol) 264.68
MDL Number MFCD03407355
InChI Key FKAIQWMEWVOGDP-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl-4-methoxybenzoate, methyl 3-chlorosulfonyl-4methoxybenzoate, 2-chlorosulphonyl-4-methoxycarbonyl anisole, methyl 3-chlorosulfonyl-4-methyloxy benzoate, methyl-3-chlorosulfonyl-4-methoxybenzoate, 2-methoxy-5-methoxycarbonyl benzenesulphonyl chloride, benzoic acid,3-chlorosulfonyl-4-methoxy-, methyl ester
PubChem CID 2794613
SMILES COC(=O)C1=CC(=C(OC)C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH3OC6H3(C(O)OCH3)S(Cl)O2
Quantity 1 g
IUPAC Name methyl 3-chlorosulfonyl-4-methoxybenzoate
Formula Weight 264.68
Percent Purity 97%
Chemical Name or Material Methyl 3-(chlorosulfonyl)-4-methoxybenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.