missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Chloro-3-oxopropionate, TRC
Tekniske data
Tekniske data
| Chemical Name or Material | Methyl 3-Chloro-3-oxopropionate |
| CAS | 37517-81-0 |
| Molecular Formula | C4H5ClO3 |
| Quantity | 10 g |
| Synonym | (Carbomethoxy)acetyl chloride, (Methoxycarbonyl)acetyl chloride, 2-Methoxycarbonylacetyl chloride, 3-Chloro-3-oxopropanoic acid methyl ester, 3-Methoxy-3-oxopropanoyl chloride, Chlorocarbonylacetic acid methyl ester, Malonic acid chloride monomethyl ester, Malonic acid monomethyl ester chloride, Methyl (chlorocarbonyl)acetate, Methyl (chloroformyl)acetate, Methyl 2-(chlorocarbonyl)acetate, Methyl 2- (chloroformyl)acetate, Methyl 3-chloro-3-oxopropanoate, Methyl 3-chloro-3-oxopropionate, Methyl malonyl chloride |
| InChI Formula | InChI=1S/C4H5ClO3/c1-8-4(7)2-3(5)6/h2H2,1H3 |
| SMILES | COC(=O)CC(=O)Cl |
| IUPAC Name | methyl 3-chloro-3-oxopropanoate |
| Molecular Weight (g/mol) | 136.53 |
| Formula Weight | 135.9927 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?