missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Chloro-3-oxopropionate, TRC

Product Code. 30475118
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475118 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475118 Supplier Toronto Research Chemicals Supplier No. TRCM3167401G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Methyl 3-Chloro-3-oxopropionate
CAS 37517-81-0
Molecular Formula C4H5ClO3
Quantity 1 g
Synonym (Carbomethoxy)acetyl chloride, (Methoxycarbonyl)acetyl chloride, 2-Methoxycarbonylacetyl chloride, 3-Chloro-3-oxopropanoic acid methyl ester, 3-Methoxy-3-oxopropanoyl chloride, Chlorocarbonylacetic acid methyl ester, Malonic acid chloride monomethyl ester, Malonic acid monomethyl ester chloride, Methyl (chlorocarbonyl)acetate, Methyl (chloroformyl)acetate, Methyl 2-(chlorocarbonyl)acetate, Methyl 2- (chloroformyl)acetate, Methyl 3-chloro-3-oxopropanoate, Methyl 3-chloro-3-oxopropionate, Methyl malonyl chloride
InChI Formula InChI=1S/C4H5ClO3/c1-8-4(7)2-3(5)6/h2H2,1H3
SMILES COC(=O)CC(=O)Cl
IUPAC Name methyl 3-chloro-3-oxopropanoate
Molecular Weight (g/mol) 136.53
Formula Weight 135.9927
Nom du produit
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.