missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromobenzoate, 98%

Product Code. 10400361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10400361 25 g Glass bottle 25g
10316261 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400361 Supplier Thermo Scientific Acros Supplier No. 333650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-89-3 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-89-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00017777
InChI Key KMFJVYMFCAIRAN-UHFFFAOYSA-N
Synonym methyl m-bromobenzoate, 3-bromobenzoic acid methyl ester, benzoic acid, 3-bromo-, methyl ester, 3-bromobenzoic acid, methyl ester, methyl-3-bromobenzoate, benzoic acid, m-bromo-, methyl ester, m-bromobenzoic acid methyl ester, pubchem3961, methyl m-bromo benzoate, methyl 3-bromo-benzoate
PubChem CID 12070
IUPAC Name methyl 3-bromobenzoate
SMILES COC(=O)C1=CC(=CC=C1)Br

Specifications

Color White to Yellow
Boiling Point 139°C to 140°C (35.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 215.05
Percent Purity 98%
Physical Form Crystalline Solid or Liquid
Chemical Name or Material Methyl 3-bromobenzoate
compliance-icons
Product Identifier
  • Methyl 3-bromobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.