missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-methoxybenzoate, 98%

Product Code. p-7029793
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11382359

Brand: Thermo Scientific Alfa Aesar H27297.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35450-37-4 | C9H9BrO3 | 245.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35450-37-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00210463
InChI Key ZREVPFANJBZHEU-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybenzoic acid methyl ester, methyl3-bromo-4-methoxybenzoate, benzoic acid, 3-bromo-4-methoxy-, methyl ester, acmc-209igx, methyl 3-bromo-p-anisate, rarechem al bf 0247, methyl-3-bromo-4-methoxybenzoate, 3-bromo-p-anisic acid methyl ester, #, methyl 3-bromo-4-methyloxy benzoate
PubChem CID 611662
IUPAC Name methyl 3-bromo-4-methoxybenzoate
SMILES COC(=O)C1=CC(Br)=C(OC)C=C1

Specifications

Melting Point 95°C to 98°C
Quantity 1 g
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-4-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.