missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminothiophene-2-carboxylate, 98+%

Product Code. 11475163
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475163 100 g 100g
11455163 5 g 5g
11465163 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475163 Supplier Thermo Scientific Alfa Aesar Supplier No. A12802.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 22288-78-4 | C6H7NO2S | 157.19 g/mol

Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-aminothiophene-2-carboxylate is used in the synthesis of 4-nitro and 4-aminothienyl ureas, total synthesis of quinazolinocarboline alkaloids and in preparation of thienopyrimidinone analogs.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22288-78-4
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00009765
InChI Key TWEQNZZOOFKOER-UHFFFAOYSA-N
Synonym methyl 3-amino-2-thiophenecarboxylate, 3-aminothiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-, methyl ester, 3-amino-thiophene-2-carboxylic acid methyl ester, 3-amino-2-thiophenecarboxylic acid methyl ester, methyl 3-amino-2-thiophene carboxylate, methyl-3-amino-thiophene-2-carboxylate, 3-amino-2-carbomethoxythiophene, 3-amino-2-methoxycarbonylthiophene, methyl3-aminothiophene-2-carboxylate
PubChem CID 89652
IUPAC Name methyl 3-aminothiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C=CS1

Specifications

Melting Point 63°C to 67°C
Boiling Point 100°C to 102°C (0.1 mmHg)
Quantity 100 g
Beilstein 1365614
Solubility Information Slightly soluble in water.
Formula Weight 157.19
Percent Purity ≥98%
Chemical Name or Material Methyl 3-aminothiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.