missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5-hydroxybenzoate, 97%, Thermo Scientific™

Product Code. p-3541074
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10071094

Brand: Thermo Scientific Maybridge MO08016DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67973-80-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09263220
InChI Key DMNGQQIFOZYIRA-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxy-benzoic acid methyl ester, benzoic acid, 3-amino-5-hydroxy-, methyl ester, pubchem17212, acmc-1bgay, dmngqqifozyira-uhfffaoysa, methyl 3-amino-5-hydroxy-benzoate, 3-amino-5-methoxycarbonyl phenol, 3-hydroxy-5-methoxycarbonyl aniline, 3-amino-5-hydroxybenzoic acid methyl ester, 5-amino-3-hydroxybenzoic acid methyl ester
PubChem CID 12438277
IUPAC Name methyl 3-amino-5-hydroxybenzoate
SMILES COC(=O)C1=CC(=CC(=C1)O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-hydroxybenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.