missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-5,6-dichloropyrazine-2-carboxylate, Mikromol™

Product Code. 30050246
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050246 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050246 Supplier Mikromol Supplier No. MM0560.080025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 3-Amino-5,6-dichloropyrazine-2-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-Amino-5,6-dichloropyrazine-2-carboxylate
CAS 1458-18-0
Analyte or Component Names Methyl 3-Amino-5,6-dichloropyrazine-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C6 H5 Cl2 N3 O2
Certifications/Compliance ISO 9001
Synonym 2-Pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, Pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester (7CI,8CI,9CI), 3-Amino-5,6-dichloropyrazine-2-carboxylic acid methyl ester
InChI Formula InChI=1S/C6H5Cl2N3O2/c1-13-6(12)2-5(9)11-4(8)3(7)10-2/h1H3,(H2,9,11)
SMILES COC(=O)c1nc(Cl)c(Cl)nc1N
IUPAC Name methyl 3-amino-5,6-dichloro-pyrazine-2-carboxylate
Molecular Weight (g/mol) 222.029
Quantity 25 mg
Formula Weight 220.9759 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.