missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, ≥95%, Thermo Scientific™

Product Code. 10056724
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10056724 5 g 5g
10734203 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10056724 Supplier Thermo Scientific Maybridge Supplier No. SEW02673DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 256353-38-5
Molecular Formula C13H10F3NO2S
Molecular Weight (g/mol) 301.28
MDL Number MFCD00832742
InChI Key FYUVISRLFPIWPP-UHFFFAOYSA-N
Synonym methyl 3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl 3-amino-4-phenyl-5-trifluoromethyl-thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-phenyl-5-trifluoromethyl-, methyl ester, maybridge1_007879, methyl 3-amino-4-phenyl-5-trifuoromethyl thiophene-2carboxylate, methyl 3-azanyl-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl-3-amino-4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, 3-amino-4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester
PubChem CID 2775320
IUPAC Name methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(=C(S1)C(F)(F)F)C1=CC=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 301.29
Percent Purity 95%
Chemical Name or Material Methyl 3-amino-4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.