missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methylbenzoate, 97%

Product Code. 11478266
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478266 100 g 100g
11458266 5 g 5g
11468266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478266 Supplier Thermo Scientific Alfa Aesar Supplier No. A15348.22
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 18595-18-1 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18595-18-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00025206
InChI Key YEPWCJHMSVABPQ-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzoic acid methyl ester, methyl 3-amino-4-methyl-benzoate, benzoic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-p-toluate, methyl 4-methyl-3-aminobenzoate, 3-amino-p-toluic acid methyl ester, 3-amino-4-methyl-benzoic acid methyl ester, 3-amino-4-methyl benzoic acid methyl ester, pubchem2351, pubchem10930
PubChem CID 337778
IUPAC Name methyl 3-amino-4-methylbenzoate
SMILES COC(=O)C1=CC=C(C)C(N)=C1

Specifications

Melting Point 113°C to 117°C
Quantity 100 g
Beilstein 2088611
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-methylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.