missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 98%, Thermo Scientific™

Product Code. 15224854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15224854 5 g Glass bottle 5g
15111017 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15224854 Supplier Thermo Scientific Acros Supplier No. 450770050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 85006-31-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00051822
InChI Key YICRPERKKBDRSP-UHFFFAOYSA-N
Synonym methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 123584
IUPAC Name methyl 3-amino-4-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(C)=CS1

Specifications

Melting Point 85°C to 88°C
Flash Point >100°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 171.22
Percent Purity 98%
Chemical Name or Material Methyl 3-amino-4-methyl-2-thiophenecarboxylate
compliance-icons
Product Identifier
  • Methyl 3-amino-4-methylthiophene-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.