missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-iodobenzoate, 97%

Product Code. 11481727
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481727 5 g 5g
11471727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481727 Supplier Thermo Scientific Alfa Aesar Supplier No. B25699.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 412947-54-7 | C8H8INO2 | 277.061 g/mol

Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 412947-54-7
Molecular Formula C8H8INO2
Molecular Weight (g/mol) 277.061
MDL Number MFCD08064225
InChI Key WJEBNIVVLJEIKE-UHFFFAOYSA-N
Synonym 3-amino-4-iodobenzoic acid methyl ester, benzoic acid, 3-amino-4-iodo-, methyl ester, methyl3-amino-4-iodobenzoate, pubchem10509, acmc-209ygt, ksc495i4l, methyl-3-amino-4-iodobenzoate, methyl 3-amino-4-iodo-benzoate, 3-amino-4-iodo-benzoic acid methyl ester
PubChem CID 21628210
IUPAC Name methyl 3-amino-4-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)N

Specifications

Melting Point 134°C to 136°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 277.06
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-4-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.