missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-methylbenzoate, 96%, Thermo Scientific™

Product Code. p-7052738
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411888 1 g 1g
15421888 5 g 5g
15431888 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411888 Supplier Thermo Scientific Alfa Aesar Supplier No. H55579.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18583-89-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00211328
InChI Key ZOOQFAUFPWXUMI-UHFFFAOYSA-N
Synonym 3-amino-2-methylbenzoic acid methyl ester, methyl3-amino-2-methylbenzoate, methyl-2-methyl-3-aminobenzoate, methyl 3-amino-2-methyl-benzoate, methyl2-methyl-3-aminobenzoate, 3-amino-o-toluic acid methyl ester, benzoic acid, 3-amino-2-methyl-, methyl ester, pubchem4978, acmc-1c8sv, ksc495o4h
PubChem CID 3801025
IUPAC Name methyl 3-amino-2-methylbenzoate
SMILES CC1=C(C=CC=C1N)C(=O)OC

Specifications

Color Colorless to Brown
Density 1.146 g/mL
Boiling Point 300°C
Flash Point 104°C (219°F)
Refractive Index 1.573
Quantity 1 g
Formula Weight 165.19
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 3-amino-2-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.