missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-methylbenzoate, 96%, Thermo Scientific™

Product Code. 15421888
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18583-89-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00211328
InChI Key ZOOQFAUFPWXUMI-UHFFFAOYSA-N
Synonym 3-amino-2-methylbenzoic acid methyl ester, methyl3-amino-2-methylbenzoate, methyl-2-methyl-3-aminobenzoate, methyl 3-amino-2-methyl-benzoate, methyl2-methyl-3-aminobenzoate, 3-amino-o-toluic acid methyl ester, benzoic acid, 3-amino-2-methyl-, methyl ester, pubchem4978, acmc-1c8sv, ksc495o4h
PubChem CID 3801025
IUPAC Name methyl 3-amino-2-methylbenzoate
SMILES CC1=C(C=CC=C1N)C(=O)OC

Specifications

Color Colorless to Brown
Density 1.146 g/mL
Boiling Point 300°C
Flash Point 104°C (219°F)
Refractive Index 1.573
Quantity 5 g
Formula Weight 165.19
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 3-amino-2-methylbenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'