missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-2-hydroxybenzoate, 97%, Thermo Scientific™

Product Code. 10068089
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10068089 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10068089 Supplier Thermo Scientific Maybridge Supplier No. MAY00290DA
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 35748-34-6
Assay Percent Range 97%
Molecular Formula C8H9NO3
MDL Number MFCD09842614
Quantity 1 g
Synonym 3-amino-2-hydroxybenzoic acid methyl ester, benzoic acid, 3-amino-2-hydroxy-, methyl ester, methyl 3-aminosalicylate, methyl3-amino-2-hydroxybenzoate, 2-amino-6-methoxycarbonyl phenol, 2-hydroxy-3-methoxycarbonyl aniline, methyl-m-aminosalicylat, acmc-209wuz, methyl-3-aminosalicylate, aminohydroxy-benzoic methyl ester
InChI Key OMWQHVRUXLRZRC-UHFFFAOYSA-N
SMILES COC(=O)C1=C(O)C(N)=CC=C1
IUPAC Name methyl 3-amino-2-hydroxybenzoate
Molecular Weight (g/mol) 167.16
PubChem CID 12782444
Formula Weight 167.16
Chemical Name or Material Methyl 3-amino-2-hydroxybenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.