missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dihydroxybenzoate, 98%

Product Code. p-7022441
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11402258

Brand: Thermo Scientific Alfa Aesar A15224.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2150-44-9 | C8H8O4 | 168.15 g/mol

Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dihydroxybenzoate was used in the synthesis of cored dendrimers. It was also used in the preparation of bis(5-carbomethoxy-1,3-phenylene)-32-crown-10, a semi-rigid 32-membered ring diester crown ether. It has been used to study ribonucleotide reductase inhibiton and anti-tumor activity .

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-44-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002289
InChI Key RNVFYQUEEMZKLR-UHFFFAOYSA-N
Synonym 3,5-dihydroxybenzoic acid methyl ester, benzoic acid, 3,5-dihydroxy-, methyl ester, methyl3,5-dihydroxybenzoate, methyl .alpha.-resorcylate, alpha-resorcinol carboxylic acid methyl ester, alpha-resorcylic acid, methyl ester, .alpha.-resorcylic acid, methyl ester, .alpha.-resorcinol carboxylic acid methyl ester, pubchem21357, methyl alpha-resorcylate
PubChem CID 75076
IUPAC Name methyl 3,5-dihydroxybenzoate
SMILES COC(=O)C1=CC(O)=CC(O)=C1

Specifications

Melting Point 164°C to 168°C
Quantity 10 g
Beilstein 2091651
Solubility Information Slightly soluble in water.
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dihydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.