missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-4-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 11477026
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
74896-66-5
C9H8Br2O2
307.97
MFCD00010729
SRLAXDDPVYZYNI-UHFFFAOYSA-N
3,5-dibromo-4-methylbenzoic acid methyl ester, methyl 3,5-dibromo-p-toluate, 3,5-dibromo-p-toluic acid methyl ester, pubchem4611, methyl3,5-dibromo-4-methylbenzoate, acmc-1bm25, zerenex e/9072914, methyl 3,5-dibromo-4-methyl-benzoate, methyl 3,5-bis bromanyl-4-methyl-benzoate, 2,6-dibromo-4-methoxycarbonyl toluene
624016
methyl 3,5-dibromo-4-methylbenzoate
COC(=O)C1=CC(Br)=C(C)C(Br)=C1
This item is not returnable. View return policy
74896-66-5
C9H8Br2O2
307.97
MFCD00010729
SRLAXDDPVYZYNI-UHFFFAOYSA-N
3,5-dibromo-4-methylbenzoic acid methyl ester, methyl 3,5-dibromo-p-toluate, 3,5-dibromo-p-toluic acid methyl ester, pubchem4611, methyl3,5-dibromo-4-methylbenzoate, acmc-1bm25, zerenex e/9072914, methyl 3,5-dibromo-4-methyl-benzoate, methyl 3,5-bis bromanyl-4-methyl-benzoate, 2,6-dibromo-4-methoxycarbonyl toluene
624016
methyl 3,5-dibromo-4-methylbenzoate
COC(=O)C1=CC(Br)=C(C)C(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 90°C
Quantity 25 g
Beilstein 2580945
Formula Weight 307.98
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-4-methylbenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'