missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-4-hydroxybenzoate, 98%

Product Code. 11439722
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439722 50 g 50g
11429722 10 g 10g
11449722 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439722 Supplier Thermo Scientific Alfa Aesar Supplier No. A10799.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41727-47-3 | C8H6Br2O3 | 309.941 g/mol

Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-dibromo-4-hydroxybenzoate is used to produce 4-hydroxy-3,5-di-pyridin-2-yl-benzoic acid methyl ester at temperature of 90-100°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41727-47-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.941
MDL Number MFCD00016416
InChI Key NVGJGYZKXBLIKY-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-hydroxybenzoic acid methyl ester, 3,5-dibromo-4-hydroxy-benzoic acid methyl ester, methyl 3,5-bis bromanyl-4-oxidanyl-benzoate, acmc-20akhs, pubchem3978, methyl dibromosalicylate, rarechem al bf 0231, timtec-bb sbb012530, labotest-bb lt00455670, labotest-bb lt03332530
PubChem CID 726975
IUPAC Name methyl 3,5-dibromo-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)Br)O)Br

Specifications

Melting Point 123°C to 125°C
Quantity 50 g
Beilstein 2373854
Solubility Information Slightly soluble in water.
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.