missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate, 99%, Thermo Scientific™

Product Code. 11318615
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11318615

Brand: Thermo Scientific Alfa Aesar L12098.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 715-33-3
Molecular Formula C9H8Br2O4
Molecular Weight (g/mol) 339.97
MDL Number MFCD00082715
InChI Key OKALYLLBFOYLQB-UHFFFAOYSA-N
Synonym 3,5-dibromo-2,4-dihydroxy-6-methylbenzoic acid methyl ester, methyl3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
PubChem CID 4121774
IUPAC Name methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
SMILES COC(=O)C1=C(C)C(Br)=C(O)C(Br)=C1O

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Beilstein 2116569
Formula Weight 339.98
Percent Purity 99%
Chemical Name or Material Methyl 3,5-dibromo-2,4-dihydroxy-6-methylbenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.