missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-diaminobenzoate, 99%

Product Code. 11393026
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393026 50 g 50g
11383026 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393026 Supplier Thermo Scientific Alfa Aesar Supplier No. L10848.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1949-55-9 | C8H10N2O2 | 166.18 g/mol

Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-55-9
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00041433
InChI Key IYWLDOMAEQHKJZ-UHFFFAOYSA-N
Synonym methyl 3,5-diaminobenzenecarboxylate, 3,5-diaminobenzoic acid methyl ester, methyldiaminobenzenecarboxylate, 3,5-diamino-benzoic acid methyl ester, benzoic acid,3,5-diamino-, methyl ester
PubChem CID 7010431
IUPAC Name methyl 3,5-diaminobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)N)N

Specifications

Melting Point 131°C to 134°C
Odor Odorless
Quantity 50 g
Beilstein 2363309
Solubility Information Insoluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl 3,5-diaminobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.