missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%, Thermo Scientific™

Product Code. 15422118
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474334-89-9
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD07369792
InChI Key LPOJJZXQESTVEV-UHFFFAOYSA-N
Synonym methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
PubChem CID 16217966
IUPAC Name methyl 3-(4-methylpiperazin-1-yl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1

Specifications

Color Colorless to Yellow
Density 1.119 g/mL
Boiling Point 169°C to 172°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 1 g
Formula Weight 234.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3-(4-methyl-1-piperazinyl)benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'