missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-hydroxyphenyl)propionate, 98%

Product Code. 15431369
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15431369

Brand: Thermo Scientific Alfa Aesar H64255.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5597-50-2 | C10H12O3 | 180.203 g/mol

Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5597-50-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00071577
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
PubChem CID 79706
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 39°C to 41°C (literature)
Color White
Boiling Point 108°C (11 mmHg)
Flash Point >113°C (235°F)
Quantity 100 g
Beilstein 2211073
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
Solubility Information Soluble in chloroform and methanol.
Formula Weight 180.2
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Methyl 3-(4-hydroxyphenyl)propionate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.