missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-hydroxyphenyl)propionate, 98%

Product Code. 15421369
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421369 25 g 25g
15431369 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421369 Supplier Thermo Scientific Alfa Aesar Supplier No. H64255.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5597-50-2 | C10H12O3 | 180.203 g/mol

Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. It can be used to prepare potent and orally available G protein-coupled receptor 40 agonists as potential antidiabetic agents. May be used in the enzymatic coupling of saccharides to protein.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5597-50-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00071577
InChI Key XRAMJHXWXCMGJM-UHFFFAOYSA-N
PubChem CID 79706
IUPAC Name methyl 3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 39°C to 41°C (literature)
Color White
Boiling Point 108°C (11 mmHg)
Flash Point >113°C (235°F)
Quantity 25 g
Beilstein 2211073
Synonym methyl 3-4-hydroxyphenyl propionate, methyl 3-4-hydroxyphenyl propanoate, benzenepropanoic acid, 4-hydroxy-, methyl ester, unii-wat13au7xb, methyl3-4-hydroxyphenyl propionate, wat13au7xb, hydrocinnamic acid, p-hydroxy-, methyl ester, 3-4-hydroxyphenyl propionic acid methyl ester, methyl 3-p-hydroxyphenyl propionate, 3-4'-hydroxyphenyl-propionic acid-ome
Solubility Information Soluble in chloroform and methanol.
Formula Weight 180.2
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Methyl 3-(4-hydroxyphenyl)propionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.