missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dichlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog Number 11430491
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2905-68-2
C8H6Cl2O2
205.03
MFCD00018165
WICGATIORFSOJL-UHFFFAOYSA-N
76193
methyl 3,4-dichlorobenzoate
COC(=O)C1=CC=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
2905-68-2
C8H6Cl2O2
205.03
MFCD00018165
WICGATIORFSOJL-UHFFFAOYSA-N
76193
methyl 3,4-dichlorobenzoate
COC(=O)C1=CC=C(Cl)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 46°C
Quantity 1 g
Synonym methyl 3,4-dichlorobenzenecarboxylate, 3,4-dichlorobenzoic acid methyl ester, benzoic acid, 3,4-dichloro-, methyl ester, benzoic acid,3,4-dichloro-, methyl ester, benzoic acid,4-dichloro-, methyl ester, acmc-1ch33, methyldichlorobenzenecarboxylate, methyl 3,4-bis chloranyl benzoate
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material Methyl 3,4-dichlorobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'