missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-Trimethoxybenzoate, TRC

Product Code. 30474817
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474817 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474817 Supplier Toronto Research Chemicals Supplier No. TRCM331060500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3,4,5-Trimethoxybenzoate
CAS 1916-07-0
Purity Grade Notes HPLC
Molecular Formula C11 H14 O5
Quantity 500 mg
Synonym Methyl 3,4,5-Trimethoxybenzoate, Trimebutine Imp. C (EP), Trimethoprim Imp. H (EP), Benzoic acid, 3,4,5-trimethoxy-, methyl ester (6CI, 7CI, 8CI, 9CI, ACI), 3,4,5-Trimethoxybenzoic acid methyl ester, Methyl eudesmate, Methyl gallate trimethyl ether, Methyl tri-O-methylgallate, NSC 16955, Trimethylgallic acid methyl ester
InChI Formula InChI=1S/C11H14O5/c1-13-8-5-7(11(12)16-4)6-9(14-2)10(8)15-3/h5-6H,1-4H3
SMILES COC(=O)c1cc(OC)c(OC)c(OC)c1
IUPAC Name methyl 3,4,5-trimethoxybenzoate
Molecular Weight (g/mol) 226.23
Formula Weight 226.0841
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.