missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-Trimethoxybenzoate, Mikromol™

Product Code. 17982716
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982716 100 mg 100mg
17951686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982716 Supplier Mikromol Supplier No. MM0093.11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 3,4,5-Trimethoxybenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3,4,5-Trimethoxybenzoate
CAS 1916-07-0
Analyte or Component Names Methyl 3,4,5-Trimethoxybenzoate
Shipping Condition Room Temperature
Molecular Formula C11 H14 O5
Certifications/Compliance ISO 17025
Synonym Trimebutine Imp. C (EP), Trimethoprim Imp. H (EP), Methyl 3,4,5-Trimethoxybenzoate, Trimebutine Maleate Impurity C, Trimethoprim Impurity H
InChI Formula InChI=1S/C11H14O5/c1-13-8-5-7(11(12)16-4)6-9(14-2)10(8)15-3/h5-6H,1-4H3
SMILES COC(=O)c1cc(OC)c(OC)c(OC)c1
IUPAC Name methyl 3,4,5-trimethoxybenzoate
Molecular Weight (g/mol) 226.23
Quantity 100 mg
Formula Weight 226.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.