missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trimethoxybenzoate, 98+%

Product Code. 11403924
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403924 25 g 25g
11413924 100 g 100g
11423924 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403924 Supplier Thermo Scientific Alfa Aesar Supplier No. A11933.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1916-07-0
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00008431
InChI Key KACHFMOHOPLTNX-UHFFFAOYSA-N
Synonym trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
PubChem CID 15956
IUPAC Name methyl 3,4,5-trimethoxybenzoate
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)OC

Specifications

Melting Point 81°C to 85°C
Boiling Point 274°C to 275°C
Quantity 25 g
Beilstein 2218156
Formula Weight 226.23
Percent Purity ≥98%
Chemical Name or Material Methyl 3,4,5-trimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.