missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethoxypropionate, 96%

Product Code. 11451247
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451247 50 g 50g
11441247 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451247 Supplier Thermo Scientific Alfa Aesar Supplier No. B24200.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7424-91-1 | C6H12O4 | 148.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7424-91-1
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.158
MDL Number MFCD00010650
InChI Key SMCVPMKCDDNUCQ-UHFFFAOYSA-N
PubChem CID 81924
IUPAC Name methyl 3,3-dimethoxypropanoate
SMILES COC(CC(=O)OC)OC

Specifications

Density 1.06
Boiling Point 76°C to 77°C (20 mmHg)
Flash Point 65°C (149°F)
Linear Formula (CH3O)2CHCH2CO2CH3
Refractive Index 1.41
Quantity 50 g
Beilstein 1561517
Synonym methyl 3,3-dimethoxypropionate, 3,3-dimethoxypropionic acid methyl ester, methyl3,3-dimethoxypropionate, propanoic acid, 3,3-dimethoxy-, methyl ester, zlchem 282, pubchem20228, acmc-1bdjk, ksc490c4j, methyl 3,3-dimethoxy-propanoate, smcvpmkcddnucq-uhfffaoysa
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material Methyl 3,3-dimethoxypropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.