missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-bromophenyl)propionate, 98%

Product Code. p-7053718
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15467868

Brand: Thermo Scientific Alfa Aesar H61342.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66191-86-4 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66191-86-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953149
InChI Key NIULOVGPRAJIJM-UHFFFAOYSA-N
Synonym methyl 3-2-bromophenyl propanoate, methyl 3-2-bromophenyl propionate, benzenepropanoic acid, 2-bromo-, methyl ester, 3-2-bromo-phenyl-propionic acid methyl ester, 3-2-bromophenyl propanoic acid,methyl ester, ksc495a4p, 2-bromohydrocinnamic acid methyl ester, 3-2-bromophenyl propanoic acid methyl ester, 3-2-bromophenyl propionic acid methyl ester
PubChem CID 15712815
IUPAC Name methyl 3-(2-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1Br

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 243.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-(2-bromophenyl)propionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.