missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(2-bromophenyl)propionate, 98%

Product Code. 15477868
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477868 25 g 25g
15467868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477868 Supplier Thermo Scientific Alfa Aesar Supplier No. H61342.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66191-86-4 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66191-86-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953149
InChI Key NIULOVGPRAJIJM-UHFFFAOYSA-N
Synonym methyl 3-2-bromophenyl propanoate, methyl 3-2-bromophenyl propionate, benzenepropanoic acid, 2-bromo-, methyl ester, 3-2-bromo-phenyl-propionic acid methyl ester, 3-2-bromophenyl propanoic acid,methyl ester, ksc495a4p, 2-bromohydrocinnamic acid methyl ester, 3-2-bromophenyl propanoic acid methyl ester, 3-2-bromophenyl propionic acid methyl ester
PubChem CID 15712815
IUPAC Name methyl 3-(2-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=CC=C1Br

Specifications

Color Colorless to Yellow
Quantity 25 g
Formula Weight 243.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-(2-bromophenyl)propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.