missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester)
Specifications
Specifications
| Chemical Name or Material | Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester) |
| CAS | 52514-63-3 |
| Analyte or Component Names | Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate (Carbidopa Methyl Ester) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C11 H16 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Carbidopa Imp. E (EP), (S)-alpha-Hydrazino-3,4-dihydroxy-alpha-methyl-benzenepropanoic acid methyl ester, Methyl (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoate, Carbidopa Impurity E |
| SMILES | COC(=O)C(C)(Cc1ccc(O)c(O)c1)NN |
| Molecular Weight (g/mol) | 240.26 |
| Quantity | 25 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction